missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Propanol, ACS Reagent, ≥99.5%, Honeywell Riedel-de Haën™
44.40 GBP valid until 2024-03-31
Use promo code "21663" to get your promotional price.
Use promo code "21663" to get your promotional price.
Click to view available options
Quantity:
500 mL
1 L
2.5 L
6 x 500 mL
20 L
6 x 1 L
Packaging:
Glass Bottle
Stainless Steel Can
Description
ACS GENERAL USE SOLVENTS
- value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
- suitable for less demanding applications such as routine isocratic HPLC
- variations and decrease issues traceable to solvent contamination
- all solvents are tested to meet ACS general use specifications
Specifications
Melting Point | -90°C |
Color | Colorless |
Linear Formula | (CH3)2CHOH |
UN Number | UN1219 |
Beilstein | 635639 |
Solubility Information | Soluble in water |
Formula Weight | 60.1g/mol |
Percent Purity | ≥99.5% |
Grade | ACS Reagent |
Density | 0.785g/mL at 25°C |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'