missing translation for 'onlineSavingsMsg'
Learn More

2-Propanol, ACS, 99.5% min, Thermo Scientific Chemicals

Catalog Number p-3256820
Click to view available options
Quantity:
1 L
4 x 1 L
This item is not returnable. View return policy
67-63-0
C3H8O
60.096
MFCD00011674
KFZMGEQAYNKOFK-UHFFFAOYSA-N
isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
3776
CHEBI:17824
propan-2-ol
CC(C)O
This item is not returnable. View return policy
67-63-0
C3H8O
60.096
MFCD00011674
KFZMGEQAYNKOFK-UHFFFAOYSA-N
isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
3776
CHEBI:17824
propan-2-ol
CC(C)O

Solvent For Gums, Resins, Alkaloids

It acts as a solvent for gums, resins, alkaloids and essential oils. It is used in cosmetic lotions. Isopropyl alcohol is used as a cleansing agent for keyboards, LCD displays and laptops. It is used as a wetting agent in the fountain solution used in lithographic printing, and is often used as a solvent for french polishing shellac, which is used in cabinet making. It act as a hydride source in the Meerwein-Ponndorf-Verley reduction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point °C to 90°C
Color Colorless
Assay Percent Range 99.5%
Appearance Liquid
Beilstein 635639
Solubility Information Miscible with water,alcohol,ether,chloroform,acetone and benzene.
Formula Weight 60.1
Grade ACS
Density 0.785 g/cm3 at 20°C
Boiling Point 82°C to 83°C
Quantity 1 L
Flash Point 12°C (53°F)
Odor Alcohol-like
Physical Form Liquid
Refractive Index 1.377
Chemical Name or Material 2-Propanol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-propanol
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : isopropyl alcohol

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'