missing translation for 'onlineSavingsMsg'
Learn More

2-Propanesulfonyl chloride, 97%, Thermo Scientific Chemicals

Catalog Number 10471474
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
10147-37-2
C3H7ClO2S
142.61
DRINJBFRTLBHNF-UHFFFAOYSA-N
isopropylsulfonyl chloride, 2-propanesulfonyl chloride, isopropylsulphonyl chloride, isopropyl sulfonyl chloride, ccris 4139, 2-propanesulfonylchloride, isopropanesulfonyl chloride, iso-propylsulfonyl chloride, 2-propane sulfonyl chloride, isopropylsulphonyl choride
82408
propane-2-sulfonyl chloride
CC(C)S(=O)(=O)Cl
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
10147-37-2
C3H7ClO2S
142.61
DRINJBFRTLBHNF-UHFFFAOYSA-N
isopropylsulfonyl chloride, 2-propanesulfonyl chloride, isopropylsulphonyl chloride, isopropyl sulfonyl chloride, ccris 4139, 2-propanesulfonylchloride, isopropanesulfonyl chloride, iso-propylsulfonyl chloride, 2-propane sulfonyl chloride, isopropylsulphonyl choride
82408
propane-2-sulfonyl chloride
CC(C)S(=O)(=O)Cl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -47.0°C
Density 1.2730g/mL
Boiling Point 95.0°C (45.0 mmHg)
Flash Point 86°C
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)2CHSO2Cl
Quantity 100 g
Beilstein 04,8
Specific Gravity 1.273
Formula Weight 142.61
Percent Purity 97%
Chemical Name or Material 2-Propanesulfonyl chloride
Show More Show Less
compliance-icons
Product Identifier
  • 2-Propanesulfonyl chloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'