Learn More
Cole-Parmer™ Standard Two-Prong Extension Clamps
Securely hold burets, thermometers, beakers, and flask necks with these extension clamps. Standard Two-Prong Extension Clamps are deal for holding straight-sided apparatus. Dual adjustable clamps are offered in either stainless steel or nickel plated zinc.
Brand: Cole-Parmer™ 08021-46
Code : A0
Additional Details : Weight : 0.80000kg
Description
Choose single adjustment style for standard use where one prong or “finger” is adjusted using a wing nut, while the other prong remains stationary or for a more versatile clamp choose the dual adjustment style.- Both prongs are easily adjusted over a wider range
- Available in medium and large sizes
- Supplied with non-slip vinyl sleeves or for temperatures above 100°C, fiberglass covers
- An additional holder must be purchased in order to attach clamps to lab-frames, support stands or other apparatus
Specifications
1 Ea. | |
Nickel-plated Zinc | |
75 mm | |
Medium |
Utility Clamp | |
Ideal for holding straight-sided apparatus such as burets, thermometers, beakers, and flask necks | |
228.6 mm |