missing translation for 'onlineSavingsMsg'
Learn More

2-PMDQ, Tocris Bioscience™

Potent, selective α1 antagonist

Brand:  Tocris (Bio-Techne) 062750

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13487823

  • £539.52 / 50mg


Estimated Shipment
04-10-2018
Add to basket

Description

Description

Potent and selective α1-adrenoceptor antagonist and antihypertensive.
Specifications

Chemical Identifiers

139047-55-5
361.449
2-[(4-Phenylpiperazin-1-yl)methyl]-2, 3-dihydroimidazo[1, 2-c]quinazolin-5(6H)-one
2-[(4-phenylpiperazin-1-yl)methyl]-3,6-dihydro-2H-imidazo[1,2-c]quinazolin-5-one
C21H23N5O
ONSOQRPYFKDOEB-UHFFFAOYSA-N
6081682
C1CN(CCN1CC2CN3C(=N2)C4=CC=CC=C4NC3=O)C5=CC=CC=C5

Specifications

2-PMDQ
139047-55-5
C21H23N5O
ONSOQRPYFKDOEB-UHFFFAOYSA-N
2-[(4-phenylpiperazin-1-yl)methyl]-3,6-dihydro-2H-imidazo[1,2-c]quinazolin-5-one
6081682
50mg
>98%
2-[(4-Phenylpiperazin-1-yl)methyl]-2, 3-dihydroimidazo[1, 2-c]quinazolin-5(6H)-one
C1CN(CCN1CC2CN3C(=N2)C4=CC=CC=C4NC3=O)C5=CC=CC=C5
361.449
361.45
Videos