missing translation for 'onlineSavingsMsg'
Learn More

2-piperidinoaniline, 97%, Thermo Scientific™

Catalog Number p-2698765
Click to view available options
Quantity:
1 g
10 g
25 g
This item is not returnable. View return policy
39643-31-7
C11H16N2
176.26
MFCD00047467
OYECAJPUPWFCSL-UHFFFAOYSA-N
2-piperidinoaniline, 2-piperidin-1-yl aniline, 2-piperidin-1-yl-phenylamine, benzenamine, 2-1-piperidinyl, n-2-aminophenyl piperidine, 2-1-piperidinyl aniline, 2-1-piperidino aniline, 1-2-aminophenyl piperidine, 2-piperidylphenylamine, 2-piperdinoaniline
458795
2-piperidin-1-ylaniline
NC1=CC=CC=C1N1CCCCC1
This item is not returnable. View return policy
39643-31-7
C11H16N2
176.26
MFCD00047467
OYECAJPUPWFCSL-UHFFFAOYSA-N
2-piperidinoaniline, 2-piperidin-1-yl aniline, 2-piperidin-1-yl-phenylamine, benzenamine, 2-1-piperidinyl, n-2-aminophenyl piperidine, 2-1-piperidinyl aniline, 2-1-piperidino aniline, 1-2-aminophenyl piperidine, 2-piperidylphenylamine, 2-piperdinoaniline
458795
2-piperidin-1-ylaniline
NC1=CC=CC=C1N1CCCCC1
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 176.26
Percent Purity 97%
Chemical Name or Material 2-Piperidinoaniline
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'