missing translation for 'onlineSavingsMsg'
Learn More

2-Piperidinemethanol, 97%, ACROS Organics™

Brand:  Acros Organics 168030250

Code : 29

Additional Details : CAS Number : 3433-37-2 Weight : 0.07500kg


Quantity 25g
Packaging Glass bottle

 View more versions of this product

Product Code. 11319736

  • £136.40 / 25g


Estimated Shipment
13-02-2018
Add to basket


Specifications

Chemical Identifiers

2-Piperidinemethanol
96.0
97%
C6H13NO
2-(Hydroxymethyl)piperidine
3433-37-2
100.0
97%
115.17

Specifications

2-Piperidinemethanol
White to pale yellow
25g
96.0
Authentic
Solid
97%
115.17
100.0° to 102.0°C (8.0 hPa)
58.0° to 64.0°C
3433-37-2
100.0
Glass bottle
97%
C6H13NO
2-(Hydroxymethyl)piperidine
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 2-Piperidinemethanol

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

SDS