missing translation for 'onlineSavingsMsg'
Learn More

2-Phosphonopropionic acid, 98+%, Thermo Scientific™

Catalog Number p-7045684
Click to view available options
Quantity:
2 g
10 g
This item is not returnable. View return policy
5962-41-4
C3H7O5P
154.058
MFCD00041415
GUXRZQZCNOHHDO-UHFFFAOYSA-N
73187
2-phosphonopropanoic acid
CC(C(=O)O)P(=O)(O)O
This item is not returnable. View return policy
5962-41-4
C3H7O5P
154.058
MFCD00041415
GUXRZQZCNOHHDO-UHFFFAOYSA-N
73187
2-phosphonopropanoic acid
CC(C(=O)O)P(=O)(O)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 118°C to 122°C
Odor Odorless
Quantity 2 g
UN Number UN3261
Beilstein 1772834
Synonym 2-phosphonopropionic acid, acmc-20annb, phosphonopropionic acid, propanoic acid,2-phosphono, propanoic acid, 2-phosphono
Formula Weight 154.06
Percent Purity ≥98%
Chemical Name or Material 2-Phosphonopropionic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'