missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(Phenylthio)ethanethioamide, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
25 g
Specifications
CAS Min % | 95.0 |
Assay Percent Range | 95% |
Packaging | Amber glass bottle |
Linear Formula | C6H5SCH2CSNH2 |
Quantity | 1 g |
IUPAC Name | 2-phenylsulfanylethanethioamide |
Formula Weight | 183.3 |
Percent Purity | 95% |
Chemical Name or Material | 2-(Phenylthio)ethanethioamide |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'