missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-(Phenylsulfonyl)thiophene, 98%

Brand:  Alfa Aesar™ A10769.06

Code : NEW

Additional Details : CAS Number : 22407-40-5 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 11429672

  • £153.80 / 5g


Estimated Shipment date
from Supplier 06-06-2018
Add to basket

Specifications

Chemical Identifiers

2- (Phenylsulfonyl)thiophene
C10H8O2S2
MFCD00052142
2-(Benzenesulfonyl)thiophene; Phenyl 2-thienyl sulfone
22407-40-5
224.3
146279

Specifications

2- (Phenylsulfonyl)thiophene
22407-40-5
C10H8O2S2
MFCD00052142
2-(Benzenesulfonyl)thiophene; Phenyl 2-thienyl sulfone
5g
98%
224.3
146279
121° to 125°C
Videos
SDS