missing translation for 'onlineSavingsMsg'
Learn More

2-Phenylpyridine, 97%, Thermo Scientific Chemicals

Catalog Number p-3593266
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
1008-89-5
C11H9N
155.20
MFCD00006280,MFCD31699954
VQGHOUODWALEFC-UHFFFAOYSA-N
pyridine, 2-phenyl, o-phenylpyridine, pyridine, phenyl, 2-phenyl-pyridine, phenyl pyridine, 2-azabiphenyl, 2-phenyl pyridine, 2-phenylpyridine, pubchem13085, 2-phenylpyridine????
13887
2-phenylpyridine
C1=CC=C(C=C1)C1=CC=CC=N1
This item is not returnable. View return policy
1008-89-5
C11H9N
155.20
MFCD00006280,MFCD31699954
VQGHOUODWALEFC-UHFFFAOYSA-N
pyridine, 2-phenyl, o-phenylpyridine, pyridine, phenyl, 2-phenyl-pyridine, phenyl pyridine, 2-azabiphenyl, 2-phenyl pyridine, 2-phenylpyridine, pubchem13085, 2-phenylpyridine????
13887
2-phenylpyridine
C1=CC=C(C=C1)C1=CC=CC=N1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.0860g/mL
Boiling Point 268°C to 270°C
Flash Point 110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula (C5H4N)C6H5
Refractive Index 1.6220 to 1.624
Quantity 5 g
Beilstein 20,424
Specific Gravity 1.086
Solubility Information Solubility in water: slightly soluble. Other solubilities: miscible in ethanol,ethyl ether
Formula Weight 155.2
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2-Phenylpyridine
Show More Show Less
compliance-icons
Product Identifier
  • 2-Phenylpyridine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'