Learn More
2-Phenylmelatonin, Tocris Bioscience™
Potent melatonin agonist
Brand:  Tocris (Bio-Techne) 068010
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 10mg
 View more versions of this product
Product Code. 13487913
  • £133.00 / 10mg
Estimated Shipment
07-03-2018
Add to basket
Description
Description
Highly potent melatonin agonist; displays higher affinity and greater potency than melatonin itself. The EC50 values for G protein activation in MT1 and MT2-transfected cells are 65 and 58 pM respectively.
Specifications
Chemical Identifiers
2-Phenylmelatonin
C19H20N2O2
308.38
N-[2-(5-Methoxy-2-phenylindol-3-yl)ethyl]acetamide
Specifications
2-Phenylmelatonin
151889-03-1
C19H20N2O2
N-[2-(5-Methoxy-2-phenylindol-3-yl)ethyl]acetamide
10mg
>98%
308.38
Videos