missing translation for 'onlineSavingsMsg'
Learn More

2-Phenylbenzoxazole, 99%, Thermo Scientific Chemicals

Catalog Number 11491377
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
833-50-1
C13H9NO
195.221
MFCD00012183
FIISKTXZUZBTRC-UHFFFAOYSA-N
2-phenylbenzoxazole, 2-phenylbenzo d oxazole, benzoxazole, 2-phenyl, ccris 7874, phenyl-2 benzoxazole french, 2-phenyl-benzoxazole, phenyl-2 benzoxazole, 2-phenybenzoxazole, 2-phenyl benzoxazole, 2-phenyl-benzooxazole
70030
2-phenyl-1,3-benzoxazole
C1=CC=C(C=C1)C2=NC3=CC=CC=C3O2
This item is not returnable. View return policy
833-50-1
C13H9NO
195.221
MFCD00012183
FIISKTXZUZBTRC-UHFFFAOYSA-N
2-phenylbenzoxazole, 2-phenylbenzo d oxazole, benzoxazole, 2-phenyl, ccris 7874, phenyl-2 benzoxazole french, 2-phenyl-benzoxazole, phenyl-2 benzoxazole, 2-phenybenzoxazole, 2-phenyl benzoxazole, 2-phenyl-benzooxazole
70030
2-phenyl-1,3-benzoxazole
C1=CC=C(C=C1)C2=NC3=CC=CC=C3O2

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 102°C to 104°C
Quantity 1 g
Formula Weight 195.22
Percent Purity 99%
Chemical Name or Material 2-Phenylbenzoxazole

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'