missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Phenyl-N-(5,6,7,8-tetrahydro-1-naphthyl)acetamide, 95%

Brand:  Alfa Aesar™ H65953.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15480037

  • £126.14 / 1g


Estimated Shipment date
from Supplier 31-01-2018
Add to basket

Specifications

Chemical Identifiers

2-Phenyl-N-(5,6,7,8-tetrahydro-1-naphthyl)acetamide
265.36
MFCD00684205

Specifications

2-Phenyl-N-(5,6,7,8-tetrahydro-1-naphthyl)acetamide
Solid
265.36
1g
95%
MFCD00684205
Videos
SDS