missing translation for 'onlineSavingsMsg'
Learn More

2-Phenyl-2-imidazoline, 98+%, ACROS Organics™

Brand:  Acros Organics 199731000

Code : 29

Additional Details : CAS Number : 936-49-2 Weight : 0.15000kg Transport : UN number : 2811 Chem class : 6.1 Pack group : III


Quantity 100g
Packaging Glass bottle

 View more versions of this product

Product Code. 10773581

  • £23.90 / 100g


Estimated Shipment
15-06-2018
Add to basket


Specifications

Chemical Identifiers

2-Phenyl-2-imidazoline
936-49-2
100.0
98+%
146.19
98+%
98.0
≥98.0%
C9H10N2
MFCD00005180

Specifications

2-Phenyl-2-imidazoline
98° to 102°C
98+%
98.0
Authentic
Flakes or Crystalline Powder
98+%
146.19
23, V,6, 425
Light yellow to yellow
100g
936-49-2
100.0
Glass bottle
≥98.0%
C9H10N2
MFCD00005180
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 2-Phenyl-2-imidazoline

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS