missing translation for 'onlineSavingsMsg'
Learn More

2-Phenyl-2-imidazoline, 98+%, Thermo Scientific™

Catalog Number 10674712
Click to view available options
Quantity:
5 g
100 g
250 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
936-49-2
C9H10N2
146.19
MFCD00005180
BKCCAYLNRIRKDJ-UHFFFAOYSA-N
2-phenyl-2-imidazoline, 2-phenylimidazoline, 1h-imidazole, 4,5-dihydro-2-phenyl, 2-imidazoline, 2-phenyl, 4,5-dihydro-2-phenyl-1h-imidazole, 2-phenyl imidazoline, 2-phenyl-2-imidazolin, acmc-209rmt, labotest-bb lt00000058, 5-23-06-00425 beilstein handbook reference
13639
2-phenyl-4,5-dihydro-1H-imidazole
C1CN=C(N1)C1=CC=CC=C1
This item is not returnable. View return policy
936-49-2
C9H10N2
146.19
MFCD00005180
BKCCAYLNRIRKDJ-UHFFFAOYSA-N
2-phenyl-2-imidazoline, 2-phenylimidazoline, 1h-imidazole, 4,5-dihydro-2-phenyl, 2-imidazoline, 2-phenyl, 4,5-dihydro-2-phenyl-1h-imidazole, 2-phenyl imidazoline, 2-phenyl-2-imidazolin, acmc-209rmt, labotest-bb lt00000058, 5-23-06-00425 beilstein handbook reference
13639
2-phenyl-4,5-dihydro-1H-imidazole
C1CN=C(N1)C1=CC=CC=C1
CAS Min % 98.0
CAS Max % 100.0
Melting Point 94°C to 99°C
Color Yellow
Boiling Point 188°C (20.0 mmHg)
Flash Point 201°C
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass bottle
Quantity 500 g
Beilstein 23, V,6, 425
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohols
Formula Weight 146.19
Percent Purity 98+%
Physical Form Crystalline Powder or Flakes
Chemical Name or Material 2-Phenyl-2-imidazoline, 98+%
Show More Show Less
compliance-icons
Product Identifier
  • 2-Phenyl-2-imidazoline
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'