missing translation for 'onlineSavingsMsg'
Learn More

2-Phenyl-1,3-thiazole-4-carbonyl chloride, Thermo Scientific™

Catalog Number p-4510196
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
36094-04-9
C10H6ClNOS
223.674
ZZFOIDMEHXGBKS-UHFFFAOYSA-N
2-phenylthiazole-4-carbonyl chloride, 2-phenyl-thiazole-4-carbonyl chloride, 4-thiazolecarbonylchloride, 2-phenyl
2776472
2-phenyl-1,3-thiazole-4-carbonyl chloride
C1=CC=C(C=C1)C2=NC(=CS2)C(=O)Cl
This item is not returnable. View return policy
36094-04-9
C10H6ClNOS
223.674
ZZFOIDMEHXGBKS-UHFFFAOYSA-N
2-phenylthiazole-4-carbonyl chloride, 2-phenyl-thiazole-4-carbonyl chloride, 4-thiazolecarbonylchloride, 2-phenyl
2776472
2-phenyl-1,3-thiazole-4-carbonyl chloride
C1=CC=C(C=C1)C2=NC(=CS2)C(=O)Cl
Assay Percent Range 95%
Quantity 250 mg
Formula Weight 223.68
Percent Purity 95%
Chemical Name or Material 2-Phenyl-1, 3-thiazole-4-carbonyl chloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'