missing translation for 'onlineSavingsMsg'
Learn More

2-Phenoxynicotinic acid, 98%, Thermo Scientific Chemicals

Catalog Number p-7081325
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
35620-71-4
C12H9NO3
215.208
MFCD00014629
CQGAXJGXGLVFGJ-UHFFFAOYSA-N
2-phenoxynicotinic acid, 3-pyridinecarboxylic acid, 2-phenoxy, 2-phenoxy-3-pyridinecarboxylic acid, 2-phenoxy-nicotinic acid, acide 2-phenoxy nicotinique french, acide 2-phenoxy nicotinique, acmc-20a2ql, maybridge1_000744, cambridge id 5832716, 2-phenoxy nicotinic acid
123450
2-phenoxypyridine-3-carboxylic acid
C1=CC=C(C=C1)OC2=C(C=CC=N2)C(=O)O
This item is not returnable. View return policy
35620-71-4
C12H9NO3
215.208
MFCD00014629
CQGAXJGXGLVFGJ-UHFFFAOYSA-N
2-phenoxynicotinic acid, 3-pyridinecarboxylic acid, 2-phenoxy, 2-phenoxy-3-pyridinecarboxylic acid, 2-phenoxy-nicotinic acid, acide 2-phenoxy nicotinique french, acide 2-phenoxy nicotinique, acmc-20a2ql, maybridge1_000744, cambridge id 5832716, 2-phenoxy nicotinic acid
123450
2-phenoxypyridine-3-carboxylic acid
C1=CC=C(C=C1)OC2=C(C=CC=N2)C(=O)O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 179°C to 182°C
Quantity 1 g
Beilstein 172906
Formula Weight 215.21
Percent Purity 98%
Chemical Name or Material 2-Phenoxynicotinic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'