missing translation for 'onlineSavingsMsg'
Learn More

2-(Phenoxymethyl)benzylamine, 97%, Maybridge

Brand:  Maybridge CC58513EA

Code : 31

Additional Details : CAS Number : 168971-56-0 Weight : 0.06000kg Transport : UN number : 2735 Chem class : 1b Pack group : III

Quantity 10g

 View more versions of this product

Product Code. 10397444

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Specifications

Chemical Identifiers

2-(Phenoxymethyl)benzylamine
96.5
97%
C14H15NO
MFCD04108110
168971-56-0
100.0
97%
213.28

Specifications

2-(Phenoxymethyl)benzylamine
168971-56-0
100.0
97%
213.28
10g
96.5
97%
C14H15NO
MFCD04108110
Videos
SDS