missing translation for 'onlineSavingsMsg'
Learn More

2-(Phenoxymethyl)benzylamine, 97%, Maybridge

Brand:  Maybridge CC58513DA

Code : 31

Additional Details : CAS Number : 168971-56-0 Weight : 0.05100kg Transport : UN number : 2735 Chem class : 1b Pack group : III

Quantity 1g
Packaging Amber Glass Bottle

 View more versions of this product

Product Code. 10144244

  • £131.00 / 1g


Estimated Shipment date
from Supplier 27-02-2018
Add to basket

Specifications

Chemical Identifiers

2-(Phenoxymethyl)benzylamine
96.5
97%
C14H15NO
MFCD04108110
168971-56-0
100.0
97%
213.28

Specifications

2-(Phenoxymethyl)benzylamine
168971-56-0
100.0
97%
C14H15NO
MFCD04108110
1g
96.5
Amber Glass Bottle
97%
213.28
Videos
SDS