missing translation for 'onlineSavingsMsg'
Learn More

2-Phenoxyethanol, 99%, Thermo Scientific Chemicals

Catalog Number p-3737182
21.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.


Click to view available options
Quantity:
250 mL
1 L
2.5 L
5 L
25 L
Packaging:
Glass bottle
Metal drum
Plastic drum
122-99-6
C8H10O2
138.17
MFCD00002857
QCDWFXQBSFUVSP-UHFFFAOYSA-N
phenoxyethanol, ethylene glycol monophenyl ether, phenyl cellosolve, ethanol, 2-phenoxy, phenoxytol, phenoxethol, phenoxetol, ethylene glycol phenyl ether, phenoxyethyl alcohol, 1-hydroxy-2-phenoxyethane
31236
CHEBI:64275
2-phenoxyethanol
C1=CC=C(C=C1)OCCO
122-99-6
C8H10O2
138.17
MFCD00002857
QCDWFXQBSFUVSP-UHFFFAOYSA-N
phenoxyethanol, ethylene glycol monophenyl ether, phenyl cellosolve, ethanol, 2-phenoxy, phenoxytol, phenoxethol, phenoxetol, ethylene glycol phenyl ether, phenoxyethyl alcohol, 1-hydroxy-2-phenoxyethane
31236
CHEBI:64275
2-phenoxyethanol
C1=CC=C(C=C1)OCCO

2-Phenoxyethanol, 99%, C8H10O2, CAS Number-122-99-6, ethylene glycol monophenyl ether, ethanol, 2-phenoxy, 1-hydroxy-2-phenoxyethane, phenoxyethanol, phenoxyethyl alcohol, phenoxethol, phenoxytol, phenoxetol, phenyl cellosolve, ethylene glycol phenyl ether, 25L, 98.5% min. (GC), 06,146, 237.0 deg.C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 11.0°C to 13.0°C
Color Colorless
Density 1.1000g/mL
Boiling Point 237.0°C
Flash Point 130°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H5OCH2CH2OH
Refractive Index 1.5360 to 1.5400
Quantity 250 mL
Beilstein 06,146
Merck Index 15,7369
Specific Gravity 1.1
Solubility Information Solubility in water: 30g/L (20°C). Other solubilities: freely soluble in alcohol and ether,miscible with acetone and ethyl acetate
Viscosity 29 mPa.s (20°C)
Formula Weight 138.17
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2-Phenoxyethanol
Show More Show Less
compliance-icons
Product Identifier
  • 2-Phenoxyethanol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H302-Harmful if swallowed.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'