missing translation for 'onlineSavingsMsg'
Learn More

2-Phenoxybenzoyl chloride, TECH, Thermo Scientific™

Catalog Number p-3175899
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
40501-36-8
C13H9ClO2
232.663
MFCD03424711
BMGKQFRMINVVPP-UHFFFAOYSA-N
benzoylchloride, 2-phenoxy, phenoxybenzoyl chloride, 2-phenoxy benzoyl chloride, 2-phenoxy-benzoyl chloride, benzoyl chloride, 2-phenoxy, 2-phenoxybenzoic acid chloride, 2-chlorocarbonyl diphenyl ether
11075303
2-phenoxybenzoyl chloride
C1=CC=C(C=C1)OC2=CC=CC=C2C(=O)Cl
This item is not returnable. View return policy
40501-36-8
C13H9ClO2
232.663
MFCD03424711
BMGKQFRMINVVPP-UHFFFAOYSA-N
benzoylchloride, 2-phenoxy, phenoxybenzoyl chloride, 2-phenoxy benzoyl chloride, 2-phenoxy-benzoyl chloride, benzoyl chloride, 2-phenoxy, 2-phenoxybenzoic acid chloride, 2-chlorocarbonyl diphenyl ether
11075303
2-phenoxybenzoyl chloride
C1=CC=C(C=C1)OC2=CC=CC=C2C(=O)Cl
CAS Min % 89.5
CAS Max % 100.0
Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 232.67
Percent Purity 90%
Chemical Name or Material 2-Phenoxybenzoyl chloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'