missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(±)-2-Pentanol, 99%, Thermo Scientific Chemicals
32.40 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
500 mL
2500 mL
Description
It finds its application as a solvent for paints and lacquers and to make drugs, to make catalyst systems and methanol and as a solvent for primers and for extracting 2-hydroxy-4-methylthiobutyric acid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It finds its application as a solvent for paints and lacquers and to make drugs, to make catalyst systems and methanol and as a solvent for primers and for extracting 2-hydroxy-4-methylthiobutyric acid.
Solubility
Soluble in water at 20°C, 166g/L. Soluble in alcohol.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
It finds its application as a solvent for paints and lacquers and to make drugs, to make catalyst systems and methanol and as a solvent for primers and for extracting 2-hydroxy-4-methylthiobutyric acid.
Solubility
Soluble in water at 20°C, 166g/L. Soluble in alcohol.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
Melting Point | °C to 50°C |
Density | 0.811 |
Boiling Point | 114°C to 115°C |
Flash Point | 33°C (91°F) |
Assay Percent Range | 0.99 |
Refractive Index | 1.406 |
Quantity | 500 mL |
UN Number | UN1105 |
Beilstein | 1718819 |
Merck Index | 14,7119 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'