missing translation for 'onlineSavingsMsg'
Learn More

2-(p-Toluenesulfonylamino)benzeneboronic acid pinacol ester, 97%, Thermo Scientific™

Catalog Number p-7029752
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
796061-07-9
C19H24BNO4S
373.274
MFCD05663862
XJERZPFENBAXMZ-UHFFFAOYSA-N
2-toluene-4-sulfonylamino phenylboronic acid, pinacol ester, 4-methyl-n-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl benzenesulfonamide, 2-p-toluenesulfonylamino benzeneboronic acid pinacol ester, 2-p-toluenesulfonylamino phenylboronic acid pinacol ester, 2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-n-tosylbenzenamine, n-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl-p-toluenesulfonamide, 2-toluene-4-sulfonylamino phenylboronic acid,pinacol ester, 4-methyl-n-2-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl phenyl benzenesulfonamide, 4-methyl-n-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl benzene-1-sulfonamide
16217718
4-methyl-N-[2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]benzenesulfonamide
B1(OC(C(O1)(C)C)(C)C)C2=CC=CC=C2NS(=O)(=O)C3=CC=C(C=C3)C
This item is not returnable. View return policy
796061-07-9
C19H24BNO4S
373.274
MFCD05663862
XJERZPFENBAXMZ-UHFFFAOYSA-N
2-toluene-4-sulfonylamino phenylboronic acid, pinacol ester, 4-methyl-n-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl benzenesulfonamide, 2-p-toluenesulfonylamino benzeneboronic acid pinacol ester, 2-p-toluenesulfonylamino phenylboronic acid pinacol ester, 2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-n-tosylbenzenamine, n-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl-p-toluenesulfonamide, 2-toluene-4-sulfonylamino phenylboronic acid,pinacol ester, 4-methyl-n-2-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl phenyl benzenesulfonamide, 4-methyl-n-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl benzene-1-sulfonamide
16217718
4-methyl-N-[2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]benzenesulfonamide
B1(OC(C(O1)(C)C)(C)C)C2=CC=CC=C2NS(=O)(=O)C3=CC=C(C=C3)C

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 135°C to 139°C
Quantity 1 g
Formula Weight 373.27
Percent Purity 97%
Chemical Name or Material 2-(p-Toluenesulfonylamino)benzeneboronic acid pinacol ester
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'