missing translation for 'onlineSavingsMsg'
Learn More

2-Oxoindoline-5-sulfonyl chloride, 97%, Thermo Scientific™

Catalog Number p-3485256
Click to view available options
Quantity:
250 mg
1 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
199328-31-9
C8H6ClNO3S
231.65
MFCD06655924
FYBNVKMJOOYPGI-UHFFFAOYSA-N
2-oxoindoline-5-sulfonyl chloride, 2-oxo-2,3-dihydro-1h-indole-5-sulfonyl chloride, 2-oxoindoline-5-sulfonylchloride, 2-oxoindoline-5-sulphonyl chloride, 2,3-dihydro-2-oxo-1h-indole-5-sulphonyl chloride, 1h-indole-5-sulfonyl chloride, 2,3-dihydro-2-oxo, 5-chlorosulfonyl indolin-2-one, 5-chlorosulphonyloxindole, 5-chlorosulfonyl oxindole, 5-chlorosufonyl-2-oxindole
4962506
ClS(=O)(=O)C1=CC2=C(NC(=O)C2)C=C1
This item is not returnable. View return policy
199328-31-9
C8H6ClNO3S
231.65
MFCD06655924
FYBNVKMJOOYPGI-UHFFFAOYSA-N
2-oxoindoline-5-sulfonyl chloride, 2-oxo-2,3-dihydro-1h-indole-5-sulfonyl chloride, 2-oxoindoline-5-sulfonylchloride, 2-oxoindoline-5-sulphonyl chloride, 2,3-dihydro-2-oxo-1h-indole-5-sulphonyl chloride, 1h-indole-5-sulfonyl chloride, 2,3-dihydro-2-oxo, 5-chlorosulfonyl indolin-2-one, 5-chlorosulphonyloxindole, 5-chlorosulfonyl oxindole, 5-chlorosufonyl-2-oxindole
4962506
ClS(=O)(=O)C1=CC2=C(NC(=O)C2)C=C1
CAS Min % 80.0
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
IUPAC Name 2-oxo-1,3-dihydroindole-5-sulfonyl chloride
Formula Weight 231.66
Percent Purity Tech.
Grade Technical
Chemical Name or Material 2-Oxoindoline-5-sulfonyl chloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'