missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Oxetanemethanol, 96%, Thermo Scientific Chemicals
53.20 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
250 mg
1 g
Description
2-Oxetanemethanol is used to produce Toluene-4-sulfonic acid oxetan-2-ylmethyl ester. Also used in medicine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Oxetanemethanol is used to produce Toluene-4-sulfonic acid oxetan-2-ylmethyl ester. Also used in medicine.
Solubility
Slightly soluble in water.
Notes
Moisture Sensitive. Store in freezer at (-20°C). Store away form water/moisture. Store away from oxidizing agent.
2-Oxetanemethanol is used to produce Toluene-4-sulfonic acid oxetan-2-ylmethyl ester. Also used in medicine.
Solubility
Slightly soluble in water.
Notes
Moisture Sensitive. Store in freezer at (-20°C). Store away form water/moisture. Store away from oxidizing agent.
Specifications
Color | Colorless to Yellow |
Density | 1.080 g/mL |
Boiling Point | 98°C (15 mmHg) |
Refractive Index | 1.446 |
Quantity | 250 mg |
Sensitivity | Moisture sensitive |
Solubility Information | Slightly soluble in water. |
Formula Weight | 88.11 |
Percent Purity | 96% |
Physical Form | Liquid |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'