missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Octyne, 98+%

Brand:  Alfa Aesar™ A10197.30

Code : NEW

Additional Details : CAS Number : 2809-67-8 Weight : 0.25000kg Transport : UN number : 3295 Chem class : 3 Pack group : III

Quantity 250g

 View more versions of this product

Product Code. 11429512

  • £445.70 / 250g


Estimated Shipment date
from Supplier 09-07-2018
Add to basket

Specifications

Chemical Identifiers

2-Octyne
CH3(CH2)4C≡CCH3
MFCD00027302
1697689
2809-67-8
110.2
UN3295
n-Pentyl methyl acetylene

Specifications

2-Octyne
≥98%
110.2
UN3295
n-Pentyl methyl acetylene
250g
Petroleum-like
2809-67-8
CH3(CH2)4C≡CCH3
MFCD00027302
1697689
137° to 138°C
28°C (82°F)
Videos
SDS