missing translation for 'onlineSavingsMsg'
Learn More

2-Octanone oxime 99% , ACROS Organics™

Brand:  Acros Organics 390281000

Code : 29

Additional Details : CAS Number : 7207-49-0 Weight : 0.15000kg Transport : UN number : 2810 Chem class : 6.1 Pack group : III


Quantity 100g
Packaging Glass bottle

 View more versions of this product

Product Code. 10772475

  • £44.00 / 100g


Estimated Shipment
15-06-2018
Add to basket


Specifications

Chemical Identifiers

2-Octanone oxime
98.5
≥98.5%
C8H17NO
143.23
7207-49-0
100.0
99%
CH3(CH2)5C(CH3)=NOH
MFCD00089167

Specifications

2-Octanone oxime
98.5
0.88g/mL
Glass bottle
≥98.5%
1.4510 to 1.4530 (20°C, 589nm)
CH3(CH2)5C(CH3)=NOH
MFCD00089167
100g
7207-49-0
100.0
Authentic
Liquid
99%
C8H17NO
143.23
Clear colorless to slightly yellow
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 2-Octanone oxime

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS