missing translation for 'onlineSavingsMsg'
Learn More

2-Octanone, 99+%, Thermo Scientific Chemicals

Catalog Number p-176735
Click to view available options
Quantity:
25 mL
500 mL
2.5 L
Packaging:
Glass bottle
This item is not returnable. View return policy
111-13-7
MFCD00009540
ZPVFWPFBNIEHGJ-UHFFFAOYSA-N
2-octanone, n-hexyl methyl ketone, hexyl methyl ketone, methyl hexyl ketone, methyl n-hexyl ketone, octanone, 2-oxooctane, 2-octanone natural, fema number 2802, unii-j2g84h29af
8093
CHEBI:87434
octan-2-one
CCCCCCC(=O)C
This item is not returnable. View return policy
111-13-7
MFCD00009540
ZPVFWPFBNIEHGJ-UHFFFAOYSA-N
2-octanone, n-hexyl methyl ketone, hexyl methyl ketone, methyl hexyl ketone, methyl n-hexyl ketone, octanone, 2-oxooctane, 2-octanone natural, fema number 2802, unii-j2g84h29af
8093
CHEBI:87434
octan-2-one
CCCCCCC(=O)C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 99.0
CAS Max % 100.0
Melting Point -16°C
Color Colorless to Yellow
Density 0.8190g/mL
Boiling Point 173°C
Flash Point 55°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Molecular Formula C8H16O
Linear Formula CH3(CH2)5COCH3
Refractive Index 1.4140 to 1.416
Quantity 25 mL
Beilstein 01, 704
Merck Index 15, 4747
Specific Gravity 0.819
Solubility Information Solubility in water: 0.9g/L. Other solubilities: miscible with alcohol,ether,hydrocarbons and,esters
Molecular Weight (g/mol) 128.21
Formula Weight 128.21
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material 2-Octanone, 99+%
Show More Show Less
compliance-icons compliance-icons
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'