missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Nitroso-1-naphthol, 97%

Brand:  Alfa Aesar™ A11980.09

Code : NEW

Additional Details : CAS Number : 132-53-6 Weight : 0.01000kg

Quantity 10g

 View more versions of this product

Product Code. 11404014

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Chemical Identifiers

2-Nitroso-1-naphthol
C10H7NO2
MFCD00003932
1-Hydroxy-2-nitrosonaphthalene
132-53-6
173.17
1867919

Specifications

2-Nitroso-1-naphthol
154° to 158°C (dec.)
10g
173.17
1867919
132-53-6
97%
C10H7NO2
MFCD00003932
1-Hydroxy-2-nitrosonaphthalene
Videos
SDS