missing translation for 'onlineSavingsMsg'
Learn More

2-Nitroethanol, 97%, Thermo Scientific Chemicals

Catalog Number p-3779302
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
625-48-9
C2H5NO3
91.07
MFCD00007405
KIPMDPDAFINLIV-UHFFFAOYSA-N
ethanol, 2-nitro, 2-nitroethan-1-ol, beta-nitroalcohol, beta-nitroethanol, 2-nitro-ethanol, .beta.-nitroethanol, ccris 6076, 2 nitroethanol, pubchem14208, 2-nitroethanol
12252
2-nitroethanol
C(CO)[N+](=O)[O-]
This item is not returnable. View return policy
625-48-9
C2H5NO3
91.07
MFCD00007405
KIPMDPDAFINLIV-UHFFFAOYSA-N
ethanol, 2-nitro, 2-nitroethan-1-ol, beta-nitroalcohol, beta-nitroethanol, 2-nitro-ethanol, .beta.-nitroethanol, ccris 6076, 2 nitroethanol, pubchem14208, 2-nitroethanol
12252
2-nitroethanol
C(CO)[N+](=O)[O-]

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point -80.0°C
Color Yellow
Density 1.2920g/mL
Boiling Point 203.0°C to 206.0°C
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula O2NCH2CH2OH
Refractive Index 1.4445
Quantity 5 g
Specific Gravity 1.292
Solubility Information Solubility in water: miscible
Formula Weight 91.07
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2-Nitroethanol
Show More Show Less
compliance-icons
Product Identifier
  • 2-Nitroethanol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'