missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Nitrobenzyl chloride, 98+%, Thermo Scientific™
Click to view available options
Quantity:
5 g
25 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
2-Nitrobenzyl chloride has been used to introduce the photo labile 2-nitrobenzyl protecting group for the phenolic OH group of tyrosine. It is used as a pharmaceutical intermediate.
Solubility
Soluble in acetone, alcohol benzene, diethyl ether, and ethanol (98%). Insoluble in water.
Notes
Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
2-Nitrobenzyl chloride has been used to introduce the photo labile 2-nitrobenzyl protecting group for the phenolic OH group of tyrosine. It is used as a pharmaceutical intermediate.
Solubility
Soluble in acetone, alcohol benzene, diethyl ether, and ethanol (98%). Insoluble in water.
Notes
Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
Specifications
Melting Point | 47°C to 51°C |
Boiling Point | 127°C to 133°C (10 mmHg) |
Flash Point | >110°C (230°F) |
Quantity | 5 g |
UN Number | UN3261 |
Beilstein | 388396 |
Formula Weight | 171.58 |
Percent Purity | ≥98% |
Chemical Name or Material | 2-Nitrobenzyl chloride |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'