missing translation for 'onlineSavingsMsg'
Learn More

2-Nitrobenzyl alcohol, 97%, Thermo Scientific Chemicals

Catalog Number p-172825
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass Bottle
This item is not returnable. View return policy
612-25-9
C7H7NO3
153.14
MFCD00007186
BWRBVBFLFQKBPT-UHFFFAOYSA-N
2-nitrobenzyl alcohol, 2-nitrophenyl methanol, o-nitrobenzyl alcohol, benzyl alcohol, o-nitro, benzenemethanol, 2-nitro, 2-nitrobenzenemethanol, o-hydroxymethylnitrobenzene, ccris 7967, benzenemethanol, nitro, 2-nitrophenyl methan-1-ol
11923
(2-nitrophenyl)methanol
OCC1=CC=CC=C1[N+]([O-])=O
This item is not returnable. View return policy
612-25-9
C7H7NO3
153.14
MFCD00007186
BWRBVBFLFQKBPT-UHFFFAOYSA-N
2-nitrobenzyl alcohol, 2-nitrophenyl methanol, o-nitrobenzyl alcohol, benzyl alcohol, o-nitro, benzenemethanol, 2-nitro, 2-nitrobenzenemethanol, o-hydroxymethylnitrobenzene, ccris 7967, benzenemethanol, nitro, 2-nitrophenyl methan-1-ol
11923
(2-nitrophenyl)methanol
OCC1=CC=CC=C1[N+]([O-])=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 69°C to 73°C
Color Beige to Beige-Brown
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass Bottle
Linear Formula O2NC6H4CH2OH
Quantity 5 g
Beilstein 06, 447
Formula Weight 153.14
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 2-Nitrobenzyl alcohol, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 2-Nitrobenzyl alcohol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'