missing translation for 'onlineSavingsMsg'
Learn More

2-Nitrobenzoyl chloride, 95%, Thermo Scientific Chemicals

Catalog Number p-3779299
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
610-14-0
C7H4ClNO3
185.57
MFCD00007131
BWWHTIHDQBHTHP-UHFFFAOYSA-N
o-nitrobenzoyl chloride, benzoyl chloride, 2-nitro, 2-nitrobenzoylchloride, benzoyl chloride, o-nitro, unii-86w2g0ja5q, nitrobenzoyl chloride, ccris 3135, 2-nitro-benzoyl chloride, benzoyl chloride, nitro, dsstox_cid_5741
11875
2-nitrobenzoyl chloride
C1=CC=C(C(=C1)C(=O)Cl)[N+](=O)[O-]
This item is not returnable. View return policy
610-14-0
C7H4ClNO3
185.57
MFCD00007131
BWWHTIHDQBHTHP-UHFFFAOYSA-N
o-nitrobenzoyl chloride, benzoyl chloride, 2-nitro, 2-nitrobenzoylchloride, benzoyl chloride, o-nitro, unii-86w2g0ja5q, nitrobenzoyl chloride, ccris 3135, 2-nitro-benzoyl chloride, benzoyl chloride, nitro, dsstox_cid_5741
11875
2-nitrobenzoyl chloride
C1=CC=C(C(=C1)C(=O)Cl)[N+](=O)[O-]

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 94.0
CAS Max % 100.0
Melting Point 17°C to 20°C
Color Brown to Yellow
Density 1.4000g/mL
Boiling Point 148°C to 149°C (9.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Linear Formula O2NC6H4COCl
Refractive Index 1.5666 to 1.5686
Quantity 5 g
Specific Gravity 1.4
Solubility Information Solubility in water: decomposes
Formula Weight 185.57
Percent Purity 95%
Physical Form Liquid After Melting
Chemical Name or Material 2-Nitrobenzoyl chloride
Show More Show Less
compliance-icons
Product Identifier
  • 2-Nitrobenzoyl chloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'