missing translation for 'onlineSavingsMsg'
Learn More

2-Nitrobenzotrifluoride, 98%, Thermo Scientific™

Catalog Number p-7077173
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
384-22-5
C7H4F3NO2
191.11
MFCD00007145
NDZJSUCUYPZXPR-UHFFFAOYSA-N
2-nitrobenzotrifluoride, 1-nitro-2-trifluoromethyl benzene, o-nitrobenzotrifluoride, 2-nitro-alpha,alpha,alpha-trifluorotoluene, o-trifluoromethyl nitrobenzene, benzene, 1-nitro-2-trifluoromethyl, unii-ovb3l8givj, ccris 2813, benzene, nitro trifluoromethyl, ovb3l8givj
9795
1-nitro-2-(trifluoromethyl)benzene
[O-][N+](=O)C1=CC=CC=C1C(F)(F)F
This item is not returnable. View return policy
384-22-5
C7H4F3NO2
191.11
MFCD00007145
NDZJSUCUYPZXPR-UHFFFAOYSA-N
2-nitrobenzotrifluoride, 1-nitro-2-trifluoromethyl benzene, o-nitrobenzotrifluoride, 2-nitro-alpha,alpha,alpha-trifluorotoluene, o-trifluoromethyl nitrobenzene, benzene, 1-nitro-2-trifluoromethyl, unii-ovb3l8givj, ccris 2813, benzene, nitro trifluoromethyl, ovb3l8givj
9795
1-nitro-2-(trifluoromethyl)benzene
[O-][N+](=O)C1=CC=CC=C1C(F)(F)F

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 31.0°C to 32.0°C
Boiling Point 216.0°C to 217.0°C
Flash Point 95°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass Bottle
Linear Formula O2NC6H4CF3
Quantity 1 g
Beilstein 05,II,251
Formula Weight 191.11
Percent Purity 98%
Chemical Name or Material 2-Nitrobenzotrifluoride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'