missing translation for 'onlineSavingsMsg'
Learn More

2-Nitrobenzoic acid, 95%, Thermo Scientific Chemicals

Catalog Number p-7080715
Click to view available options
Quantity:
100 g
500 g
2500 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
552-16-9
C7H5NO4
167.12
MFCD00007137
SLAMLWHELXOEJZ-UHFFFAOYSA-N
2-nitrobenzoic acid, o-nitrobenzoic acid, benzoic acid, 2-nitro, 2-nitrobenzoicacid, o-carboxynitrobenzene, benzoic acid, o-nitro, nitrobenzoic acid, 2-nitro-benzoic acid, ccris 2334, 2-nitro benzoic acid
11087
CHEBI:25620
C1=CC=C(C(=C1)C(=O)O)[N+](=O)[O-]
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
552-16-9
C7H5NO4
167.12
MFCD00007137
SLAMLWHELXOEJZ-UHFFFAOYSA-N
2-nitrobenzoic acid, o-nitrobenzoic acid, benzoic acid, 2-nitro, 2-nitrobenzoicacid, o-carboxynitrobenzene, benzoic acid, o-nitro, nitrobenzoic acid, 2-nitro-benzoic acid, ccris 2334, 2-nitro benzoic acid
11087
CHEBI:25620
C1=CC=C(C(=C1)C(=O)O)[N+](=O)[O-]

It is employed as a synthetic reagent and can function as a growth supplement for Pseudomonas fluorescens strain KU-7. It also finds its use as a reagent for protection of amine groups and in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a synthetic reagent and can function as a growth supplement for Pseudomonas fluorescens strain KU-7. It also finds its use as a reagent for protection of amine groups and in organic synthesis.

Solubility
Soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY
Melting Point 146°C to 149°C
Density 1.58
Quantity 100 g
Beilstein 1910632
Merck Index 14,6589
Solubility Information Soluble in water.
Formula Weight 167.12
Percent Purity 95%
Chemical Name or Material 2-Nitrobenzoic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'