missing translation for 'onlineSavingsMsg'
Learn More

2-Nitrobenzaldehyde, 97%, Thermo Scientific™

Catalog Number 10646514
Click to view available options
Quantity:
10 g
50 g
100 g
This item is not returnable. View return policy
552-89-6
C7H5NO3
151.121
MFCD00007132
CMWKITSNTDAEDT-UHFFFAOYSA-N
o-nitrobenzaldehyde, benzaldehyde, 2-nitro, nitrobenzaldehyde, benzaldehyde, o-nitro, 2-nitro-benzaldehyde, 2-formylnitrobenzene, ccris 2322, unii-48b18q9b8e, benzaldehyde, nitro, ortho-nitrobenzaldehyde
11101
CHEBI:66927
2-nitrobenzaldehyde
C1=CC=C(C(=C1)C=O)[N+](=O)[O-]
This item is not returnable. View return policy
552-89-6
C7H5NO3
151.121
MFCD00007132
CMWKITSNTDAEDT-UHFFFAOYSA-N
o-nitrobenzaldehyde, benzaldehyde, 2-nitro, nitrobenzaldehyde, benzaldehyde, o-nitro, 2-nitro-benzaldehyde, 2-formylnitrobenzene, ccris 2322, unii-48b18q9b8e, benzaldehyde, nitro, ortho-nitrobenzaldehyde
11101
CHEBI:66927
2-nitrobenzaldehyde
C1=CC=C(C(=C1)C=O)[N+](=O)[O-]
CAS Min % 97.0
Assay Percent Range 97%
Linear Formula O2NC6H4CHO
Quantity 100 g
Formula Weight 151.12
Percent Purity 97%
Chemical Name or Material 2-Nitrobenzaldehyde
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'