missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Nitro-N-n-propylbenzenesulfonamide, 97%

Brand:  Alfa Aesar™ H60481.03

Code : NEW

Additional Details : CAS Number : 89840-63-1 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15456948

  • £358.49 / 1g


Estimated Shipment date
from Supplier 30-01-2018
Add to basket

Specifications

Chemical Identifiers

2-Nitro-N-n-propylbenzenesulfonamide
244.27
3351864
89840-63-1
MFCD00514615

Specifications

2-Nitro-N-n-propylbenzenesulfonamide
61° to 62°C
89840-63-1
97%
MFCD00514615
Pale yellow
1g
Solid
244.27
3351864
Videos
SDS