Alfa Aesar™ 2-Nitro-N-(4-methylphenyl)benzenesulfonamide, 97%


Overview
Brand: Alfa Aesar™

Manufacturer Part Number: H60960.03

UNSPSC: 12352100

Code: NEW

Additional Details:
Additional Details: CAS Number: 63228-66-0 Weight: 0.00100kg


Quantity 1g

Product Code. 15467458

Quantity Price
1 £ 358.49 / 1g
Estimated Shipment date
from Supplier 11-04-2017
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material 2-Nitro-N-(4-methylphenyl)benzenesulfonamide
CAS 63228-66-0
Formula Weight 292.31
MDL Number MFCD00577610
Beilstein 3393448

Specification

Formula Weight 292.31
Assay 97%
Physical Form Solid
Chemical Name or Material 2-Nitro-N-(4-methylphenyl)benzenesulfonamide
CAS 63228-66-0
Quantity 1g
Color Red
MDL Number MFCD00577610
Beilstein 3393448
Documentation

Technical documents