missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Nitro-N-(3-nitrophenyl)benzenesulfonamide, 97%

Brand:  Alfa Aesar™ H60347.03

Code : NEW

Additional Details : CAS Number : 63228-70-6 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15466808

  • £358.49 / 1g


Estimated Shipment date
from Supplier 06-02-2018
Add to basket

Specifications

Chemical Identifiers

2-Nitro-N-(3-nitrophenyl)benzenesulfonamide
63228-70-6
323.28
MFCD00401913

Specifications

2-Nitro-N-(3-nitrophenyl)benzenesulfonamide
63228-70-6
97%
MFCD00401913
1g
Solid
323.28
Videos
SDS