missing translation for 'onlineSavingsMsg'
Learn More

2-Nitro-m-xylene, 99%, Thermo Scientific Chemicals

Catalog Number p-7044264
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy
81-20-9
C8H9NO2
151.165
MFCD00007181
HDFQKJQEWGVKCQ-UHFFFAOYSA-N
2-nitro-m-xylene, 2,6-dimethylnitrobenzene, benzene, 1,3-dimethyl-2-nitro, 2,6-dimethyl-1-nitrobenzene, m-xylene, 2-nitro, ccris 3119, 1,3-dimethyl-2-nitro-benzene, dsstox_cid_5134, dsstox_rid_77678, dsstox_gsid_25134
6672
1,3-dimethyl-2-nitrobenzene
CC1=C(C(=CC=C1)C)[N+](=O)[O-]
This item is not returnable. View return policy
81-20-9
C8H9NO2
151.165
MFCD00007181
HDFQKJQEWGVKCQ-UHFFFAOYSA-N
2-nitro-m-xylene, 2,6-dimethylnitrobenzene, benzene, 1,3-dimethyl-2-nitro, 2,6-dimethyl-1-nitrobenzene, m-xylene, 2-nitro, ccris 3119, 1,3-dimethyl-2-nitro-benzene, dsstox_cid_5134, dsstox_rid_77678, dsstox_gsid_25134
6672
1,3-dimethyl-2-nitrobenzene
CC1=C(C(=CC=C1)C)[N+](=O)[O-]

2-Nitro-m-xylene is employed as a pharmaceutical intermediate in organic synthesis. It is also used in the preparation of Direct Violet 7 and solvent Red 26. Further, it is used in the petrochemical industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Nitro-m-xylene is employed as a pharmaceutical intermediate in organic synthesis. It is also used in the preparation of Direct Violet 7 and solvent Red 26. Further, it is used in the petrochemical industry.

Solubility
Miscible with ethanol. Immiscible with water.

Notes
Incompatible with strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY
Melting Point 14°C to 16°C
Density 1.11
Boiling Point 225°C to 227°C
Flash Point 87°C (188°F)
Refractive Index 1.522
Quantity 100 g
UN Number UN1665
Beilstein 2046066
Solubility Information Miscible with ethanol. Immiscible with water.
Formula Weight 151.17
Percent Purity 99%
Chemical Name or Material 2-Nitro-m-xylene
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'