missing translation for 'onlineSavingsMsg'
Learn More

2-Nitro-4-(trifluoromethyl)phenol, 99%, ACROS Organics™

Brand:  Acros Organics 218880050

Code : 29

Additional Details : CAS Number : 400-99-7 Weight : 0.05500kg


Quantity 5g

 View more versions of this product

Product Code. 11954691

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Chemical Identifiers

2-Nitro-4-(trifluoromethyl)phenol
98.5
99%
C7H4F3NO3
207.11
400-99-7
100.0
99%
O2NC6H3(CF3)OH

Specifications

2-Nitro-4-(trifluoromethyl)phenol
98.5
Authentic
1.4700g/mL
Yellow to orange
99%
5g
C7H4F3NO3
207.11
400-99-7
100.0
1.5035
92.0° to 94.0°C (12.0 Torr)
Liquid
99%
95°C
O2NC6H3(CF3)OH
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 2-Nitro-4-(trifluoromethyl)phenol

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

SDS