missing translation for 'onlineSavingsMsg'
Learn More

2-Naphthoyl chloride, 98%, Thermo Scientific Chemicals

Catalog Number 10873961
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy
2243-83-6
C11H7ClO
190.63
MFCD00004093
XNLBCXGRQWUJLU-UHFFFAOYSA-N
2-naphthoyl chloride, 2-naphthalenecarbonyl chloride, 2-naphthoic chloride, 2-naphthoylchloride, beta-naphthoyl chloride, 2-napthoyl chloride, 2-chlorocarbonyl naphthalene, .beta.-naphthoyl chloride, .beta.-naphthalenecarbonyl chloride, beta-naphthalenecarbonyl chloride
75246
naphthalene-2-carbonyl chloride
C1=CC=C2C=C(C=CC2=C1)C(=O)Cl
This item is not returnable. View return policy
2243-83-6
C11H7ClO
190.63
MFCD00004093
XNLBCXGRQWUJLU-UHFFFAOYSA-N
2-naphthoyl chloride, 2-naphthalenecarbonyl chloride, 2-naphthoic chloride, 2-naphthoylchloride, beta-naphthoyl chloride, 2-napthoyl chloride, 2-chlorocarbonyl naphthalene, .beta.-naphthoyl chloride, .beta.-naphthalenecarbonyl chloride, beta-naphthalenecarbonyl chloride
75246
naphthalene-2-carbonyl chloride
C1=CC=C2C=C(C=CC2=C1)C(=O)Cl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 156.0°C to 158.0°C (7.0mmHg)
Melting Point 49.0°C to 54.0°C
Color Brown to Gray
Physical Form Crystalline Chunks
Quantity 50 g
Assay Percent Range 97.5% min. (GC)
Linear Formula C10H7COCl
Flash Point >110°C
Solubility Information Solubility in water: decomposes. Other solubilities: soluble in aceton
Formula Weight 190.63
Percent Purity 98%
Infrared Spectrum Authentic
Packaging Glass bottle
Chemical Name or Material 2-Naphthoyl chloride
Show More Show Less
compliance-icons
Product Identifier
  • 2-Naphthoyl chloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'