missing translation for 'onlineSavingsMsg'
Learn More

2-Naphthaldehyde, 98%, ACROS Organics™

£20.30 - £192.00

Chemical Identifiers

CAS 66-99-9
Molecular Formula C11H8O
Molecular Weight (g/mol) 156.18
MDL Number MFCD00004094
InChI Key PJKVFARRVXDXAD-UHFFFAOYSA-N
Synonym 2-naphthaldehyde, 2-naphthalenecarboxaldehyde, 2-formylnaphthalene, beta-naphthaldehyde, beta-formylnaphthalene, beta-naphthylaldehyde, beta-naphthylcarboxaldehyde, .beta.-naphthaldehyde, .beta.-naphthylaldehyde, .beta.-formylnaphthalene
PubChem CID 6201
ChEBI CHEBI:52368
IUPAC Name naphthalene-2-carbaldehyde
SMILES C1=CC=C2C=C(C=CC2=C1)C=O
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10036612
SDS View Documents
Acros Organics
128100050
5g Glass bottle
£20.30
5g
View alternative products
Save Up to 
»
In stock, ready to ship: 2
Please to purchase this item. Need a web account? Register with us today!
10503871
SDS View Documents
Acros Organics
128100250
25g Glass bottle
£65.80
25g
View alternative products
Save Up to 
»
In stock, ready to ship: 8
Please to purchase this item. Need a web account? Register with us today!
10564074
SDS View Documents
Acros Organics
128101000
100g Glass bottle
£192.00
100g
View alternative products
Save Up to 
»
Estimated Shipment: 11-10-2021
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

66-99-9
156.18
PJKVFARRVXDXAD-UHFFFAOYSA-N
6201
naphthalene-2-carbaldehyde
C11H8O
MFCD00004094
2-naphthaldehyde, 2-naphthalenecarboxaldehyde, 2-formylnaphthalene, beta-naphthaldehyde, beta-formylnaphthalene, beta-naphthylaldehyde, beta-naphthylcarboxaldehyde, .beta.-naphthaldehyde, .beta.-naphthylaldehyde, .beta.-formylnaphthalene
CHEBI:52368
C1=CC=C2C=C(C=CC2=C1)C=O

Specifications

2-Naphthaldehyde
Authentic
160.0°C (19.0 mmHg)
58.0°C to 62.0°C
66-99-9
100.0
C11H8O
MFCD00004094
2-naphthaldehyde, 2-naphthalenecarboxaldehyde, 2-formylnaphthalene, beta-naphthaldehyde, beta-formylnaphthalene, beta-naphthylaldehyde, beta-naphthylcarboxaldehyde, .beta.-naphthaldehyde, .beta.-naphthylaldehyde, .beta.-formylnaphthalene
PJKVFARRVXDXAD-UHFFFAOYSA-N
naphthalene-2-carbaldehyde
6201
156.18
98%
>100°C
Glass bottle
Beige or White to Yellow
5g
97.5
97.5% min. (GC)
C10H7CHO
07, 401
Solubility in water: slightly soluble. Other solubilities: soluble in hot water
C1=CC=C2C=C(C=CC2=C1)C=O
156.18
CHEBI:52368
Powder or Chunks
Safety and Handling

Safety and Handling

Product Identifier
  • 2-Naphthaldehyde

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Product Certifications