missing translation for 'onlineSavingsMsg'
Learn More

2-Naphthaldehyde, 98%, Thermo Scientific Chemicals

Catalog Number 10564074
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
66-99-9
C11H8O
156.18
MFCD00004094
PJKVFARRVXDXAD-UHFFFAOYSA-N
2-naphthaldehyde, 2-naphthalenecarboxaldehyde, 2-formylnaphthalene, beta-naphthaldehyde, beta-formylnaphthalene, beta-naphthylaldehyde, beta-naphthylcarboxaldehyde, .beta.-naphthaldehyde, .beta.-naphthylaldehyde, .beta.-formylnaphthalene
6201
CHEBI:52368
naphthalene-2-carbaldehyde
C1=CC=C2C=C(C=CC2=C1)C=O
66-99-9
C11H8O
156.18
MFCD00004094
PJKVFARRVXDXAD-UHFFFAOYSA-N
2-naphthaldehyde, 2-naphthalenecarboxaldehyde, 2-formylnaphthalene, beta-naphthaldehyde, beta-formylnaphthalene, beta-naphthylaldehyde, beta-naphthylcarboxaldehyde, .beta.-naphthaldehyde, .beta.-naphthylaldehyde, .beta.-formylnaphthalene
6201
CHEBI:52368
naphthalene-2-carbaldehyde
C1=CC=C2C=C(C=CC2=C1)C=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 58.0°C to 62.0°C
Color Beige or White to Yellow
Boiling Point 160.0°C (19.0 mmHg)
Flash Point >100°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula C10H7CHO
Quantity 100 g
Beilstein 07, 401
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in hot water
Formula Weight 156.18
Percent Purity 98%
Physical Form Chunks and/or Powder
Chemical Name or Material 2-Naphthaldehyde
Show More Show Less
compliance-icons
Product Identifier
  • 2-Naphthaldehyde
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'