missing translation for 'onlineSavingsMsg'
Learn More

2-n-Propylthiazole, 99%, Thermo Scientific Chemicals

Catalog Number p-7081557
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
17626-75-4
C6H9NS
127.205
MFCD00053154
CMOIEFFAOUQJPS-UHFFFAOYSA-N
87198
2-propyl-1,3-thiazole
CCCC1=NC=CS1
This item is not returnable. View return policy
17626-75-4
C6H9NS
127.205
MFCD00053154
CMOIEFFAOUQJPS-UHFFFAOYSA-N
87198
2-propyl-1,3-thiazole
CCCC1=NC=CS1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 1.03
Boiling Point 62°C (15 mmHg)
Flash Point 54°C (129°F)
Refractive Index 1.5055
Quantity 5 g
UN Number UN1993
Synonym 2-propylthiazole, thiazole, 2-propyl, 2-n-propylthiazole, unii-t395979vrl, 2-propylthiazol, thiazole,2-propyl, 2-normal-propyl thiazole, acmc-209eb6, #
Formula Weight 127.2
Percent Purity 99%
Chemical Name or Material 2-n-Propylthiazole

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'