missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-n-Propyl-1-pentanol, 97%

Brand:  Alfa Aesar™ H57954.06

Code : NEW

Additional Details : CAS Number : 58175-57-8 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15404348

  • £85.00 / 5g


Estimated Shipment date
from Supplier 24-04-2018
Add to basket

Specifications

Chemical Identifiers

2-n-Propyl-1-pentanol
(CH3CH2CH2)2CHCH2OH
MFCD00010260
58175-57-8
130.23

Specifications

2-n-Propyl-1-pentanol
80° to 82°C (12mmHg)
5g
1.4300
97%
130.23
0.83g/mL
Colorless to pale yellow
58175-57-8
Liquid
(CH3CH2CH2)2CHCH2OH
MFCD00010260
Videos
SDS