missing translation for 'onlineSavingsMsg'
Learn More

2-(morpholinosulfonyl)benzoic acid, 97%, Thermo Scientific™

Catalog Number p-3586629
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
87223-34-5
C11H13NO5S
271.287
MFCD09813541
YTQKWJNSSYODEV-UHFFFAOYSA-N
2-morpholinosulfonyl benzoic acid, 2-morpholine-4-sulfonyl benzoic acid, 2-morpholin-4-ylsulfonyl benzoic acid, benzoic acid,2-4-morpholinylsulfonyl, 2-morpholin-4-ylsulphonyl benzoic acid
13140207
2-morpholin-4-ylsulfonylbenzoic acid
C1COCCN1S(=O)(=O)C2=CC=CC=C2C(=O)O
This item is not returnable. View return policy
87223-34-5
C11H13NO5S
271.287
MFCD09813541
YTQKWJNSSYODEV-UHFFFAOYSA-N
2-morpholinosulfonyl benzoic acid, 2-morpholine-4-sulfonyl benzoic acid, 2-morpholin-4-ylsulfonyl benzoic acid, benzoic acid,2-4-morpholinylsulfonyl, 2-morpholin-4-ylsulphonyl benzoic acid
13140207
2-morpholin-4-ylsulfonylbenzoic acid
C1COCCN1S(=O)(=O)C2=CC=CC=C2C(=O)O
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 271.29
Percent Purity 97%
Chemical Name or Material 2-(Morpholinosulfonyl)benzoic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'