missing translation for 'onlineSavingsMsg'
Learn More

(2-Morpholinopyrid-4-yl)methylamine, 97%, Thermo Scientific™

Catalog Number 10568253
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
864068-88-2
C10H15N3O
193.25
MFCD08060472
NHFVDBCQOZATNT-UHFFFAOYSA-N
2-morpholinopyrid-4-yl methylamine, 2-morpholin-4-yl pyridin-4-yl methanamine, 2-morpholin-4-ylpyridin-4-yl methylamine, 4-pyridinemethanamine,2-4-morpholinyl, 1-2-morpholin-4-yl pyridin-4-yl methanamine, 2-morpholin-4-ylpyridin-4-yl methanamine, 2-morpholinopyridin-4-yl methanamine, 2-morpholin-4-yl-4-pyridyl methylamine, 4-pyridinemethanamine, 2-4-morpholinyl
7537568
(2-morpholin-4-ylpyridin-4-yl)methanamine
C1COCCN1C2=NC=CC(=C2)CN
This item is not returnable. View return policy
864068-88-2
C10H15N3O
193.25
MFCD08060472
NHFVDBCQOZATNT-UHFFFAOYSA-N
2-morpholinopyrid-4-yl methylamine, 2-morpholin-4-yl pyridin-4-yl methanamine, 2-morpholin-4-ylpyridin-4-yl methylamine, 4-pyridinemethanamine,2-4-morpholinyl, 1-2-morpholin-4-yl pyridin-4-yl methanamine, 2-morpholin-4-ylpyridin-4-yl methanamine, 2-morpholinopyridin-4-yl methanamine, 2-morpholin-4-yl-4-pyridyl methylamine, 4-pyridinemethanamine, 2-4-morpholinyl
7537568
(2-morpholin-4-ylpyridin-4-yl)methanamine
C1COCCN1C2=NC=CC(=C2)CN
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 193.25
Percent Purity 97%
Chemical Name or Material (2-Morpholinopyrid-4-yl)methylamine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'