missing translation for 'onlineSavingsMsg'
Learn More

(2-Morpholinophenyl)methanol, 97%, Thermo Scientific™

Catalog Number p-2695085
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
465514-33-4
C11H15NO2
193.25
MFCD03086181
MYGVYNRBQSAMIF-UHFFFAOYSA-N
2-morpholin-4-yl-phenyl methanol, 2-morpholinophenyl methanol, 2-morpholin-4-yl phenyl methanol, 2-morpholin-4-ylphenyl methanol, benzenemethanol,2-4-morpholinyl, 2-morpholinophenylmethanol, 2-morpholin-4-ylphenyl methan-1-ol
2776562
OCC1=CC=CC=C1N1CCOCC1
This item is not returnable. View return policy
465514-33-4
C11H15NO2
193.25
MFCD03086181
MYGVYNRBQSAMIF-UHFFFAOYSA-N
2-morpholin-4-yl-phenyl methanol, 2-morpholinophenyl methanol, 2-morpholin-4-yl phenyl methanol, 2-morpholin-4-ylphenyl methanol, benzenemethanol,2-4-morpholinyl, 2-morpholinophenylmethanol, 2-morpholin-4-ylphenyl methan-1-ol
2776562
OCC1=CC=CC=C1N1CCOCC1
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
IUPAC Name (2-morpholin-4-ylphenyl)methanol
Formula Weight 193.24
Percent Purity 97%
Chemical Name or Material (2-Morpholinophenyl)methanol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'